(S)-(+)-2-Phenylbutyric acid ≥99%

279889-5G 279889-1G
220051-048EA 340.31 USD
220051-048 220051-047
(S)-(+)-2-Phenylbutyric acid ≥99%
(S)-(+)-2-Phenylbutyric acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00063164
CAS Number: 4286-15-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR