Triethylsilyl trifluoromethanesulfonate ≥99%

Synonyms: Triethylsilyl trifluoromethanesulphonate

279471-50G 279471-10G
220050-961EA 253.08 USD
220050-961 220050-960
Triethylsilyl trifluoromethanesulfonate ≥99%
Triethylsilyl trifluoromethanesulphonate
Formula: C₇H₁₅F₃O₃SSi
MW: 264.34 g/mol
Boiling Pt: 77…79 °C (1013 hPa)
Density: 1.225 g/cm³ (20 °C)
MDL Number: MFCD00000407
CAS Number: 79271-56-0
EINECS: 279-124-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR