4-Amino-2-chlorobenzonitrile ≥99%

Warning

Synonyms: 3-Chloro-4-cyanoaniline

279013-50G 279013-10G
220050-936EA 385.38 USD
220050-936 220050-935
4-Amino-2-chlorobenzonitrile ≥99%
4-Amino-2-chlorobenzonitrile
Formula: C₇H₅ClN₂
MW: 152.58 g/mol
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 162…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00035926
CAS Number: 20925-27-3
EINECS: 244-114-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR