3-Iodo-p-toluic acid ≥97%

Warning

278858-5G 278858-25G
220050-923EA 480.85 USD
220050-923 220050-922
3-Iodo-p-toluic acid ≥97%
3-Iodo-p-toluic acid
Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 329 °C (1013 hPa)
Melting Pt: 210 - 211 °C
MDL Number: MFCD00010392
CAS Number: 82998-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR