4'-Hydroxyacetophenone ≥99%

278564-5G 278564-500G 278564-100G
220050-898EA 49.54 USD
220050-898 220050-897 220050-896
4'-Hydroxyacetophenone ≥99%
4'-Hydroxyacetophenone
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 105…108 °C
Density: 1.139 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002359
CAS Number: 99-93-4
EINECS: 202-802-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR