Digadolinium trioxide ≥99.9%

Warning

Synonyms: Gadolinium (III) oxide

278513-25G 278513-100G
220050-895EA 153.69 USD
220050-895 220050-894
Digadolinium trioxide ≥99.9%
Digadolinium trioxide
Formula: Gd₂O₃
MW: 362.5 g/mol
Melting Pt: 2410 °C
Density: 7.407 g/cm³ (20 °C)
MDL Number: MFCD00011026
CAS Number: 12064-62-9
EINECS: 235-060-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR