Palladium 5% on barium sulfate, reduced

277991-1G 277991-10G 277991-50G
220050-202EA 31.15 USD
220050-202 220050-886 220050-203
Palladium 5% on barium sulfate, reduced
Palladium
Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR