Ammonium molybdate(VI)

Synonyms: Molybdic acid ammonium salt, Molybdic acid, ammonium salt, Ammonium molybdate, di-Ammonium molybdate

277908-5G 277908-100G 277908-20G
220050-877EA 149.85 USD
220050-877 220050-875 220050-876
Ammonium molybdate(VI)
Ammonium molybdate(VI)
Formula: H₈MoN₂O₄
MW: 196.01 g/mol
MDL Number: MFCD00011596
CAS Number: 13106-76-8
EINECS: 236-031-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR