4-Methylvaleric acid ≥99%

Danger

Synonyms: Isocaproic acid, 4-Methylpentanoic acid

277827-25G 277827-5G
220049-709EA 106.94 USD
220049-709 220049-710
4-Methylvaleric acid ≥99%
4-Methylvaleric acid
Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: -35 °C
Density: 0.9192 g/cm³ (20 °C)
Flash Pt: 89 °C
Storage Temperature: Ambient
MDL Number: MFCD00002803
CAS Number: 646-07-1
EINECS: 211-464-0

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR