1-Bromo-3,5-dichlorobenzene ≥98%

Warning

Synonyms: 3,5-Dichlorobromobenzene

277789-100G 277789-25G
220049-706EA 110.92 USD
220049-706 220049-707
1-Bromo-3,5-dichlorobenzene ≥98%
1-Bromo-3,5-dichlorobenzene
Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 82…84 °C
MDL Number: MFCD00000584
CAS Number: 19752-55-7
EINECS: 243-270-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR