Iodomethane[¹³C]

Danger

Synonyms: Methyl iodide-(13C)

277185-1G 277185-5G
220050-830EA 250 USD
220050-830 220050-831
Iodomethane[¹³C]
Iodomethane[¹³C]
Formula: H₃CI
MW: 142.93 g/mol
MDL Number: MFCD00001075
CAS Number: 4227-95-6
UN: 2644
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR