5-Bromo-m-xylene ≥97%

Warning

Synonyms: 1-Bromo-3,5-dimethylbenzene

276316-100G 276316-25G
220050-733EA 161.54 USD
220050-733 220050-734
5-Bromo-m-xylene ≥97%
5-Bromo-m-xylene
Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 60…62 °C (5.33 hPa)
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000087
CAS Number: 556-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR