4'-Ethoxyacetophenone ≥98%

275719-25G 275719-100G
220050-201EA 150.38 USD
220050-201 220050-200
4'-Ethoxyacetophenone ≥98%
4'-Ethoxyacetophenone
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 83 °C (0.04 mmHg)
Melting Pt: 39 °C
MDL Number: MFCD00009095
CAS Number: 1676-63-7
EINECS: 216-825-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR