2,5-Dimethoxythiophenol ≥98%

Synonyms: 2,5-Dimethoxybenzenethiol, 2,5-Dimethoxyphenyl mercaptan, 1,4-Dimethoxy-2-mercaptobenzene, 2,5-Dimethoxybenzene-1-thiol

275425-1G 275425-5G
220050-188EA 50.62 USD
220050-188 220050-189
2,5-Dimethoxythiophenol ≥98%
2,5-Dimethoxythiophenol
Formula: C₈H₁₀O₂S
MW: 170.23 g/mol
MDL Number: MFCD00052738
CAS Number: 1483-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR