2-Fluorothiophenol ≥97%

Danger

Synonyms: 2-Fluorobenzenethiol, 1-Fluoro-2-mercaptobenzene, 2-Fluorobenzene-1-thiol, o-Fluorothiophenol

275379-1G 275379-5G
220050-181EA 34.15 USD
220050-181 220050-182
2-Fluorothiophenol ≥97%
2-Fluorothiophenol
Formula: C₆H₅FS
MW: 128.17 g/mol
Boiling Pt: 162 - 164 °C (1013 hPa)
Density: 1.191 g/cm³ (30 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00041419
CAS Number: 2557-78-0
UN: 3071
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR