9,10-Phenanthrenequinone

Synonyms: Phenanthrene-9,10-dione

275034-25G 275034-100G
220050-593EA 133.38 USD
220050-593 220050-592
9,10-Phenanthrenequinone
9,10-Phenanthrenequinone
Formula: C₁₄H₈O₂
MW: 208.22 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 203…206 °C
Density: 1.4045 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001163
CAS Number: 84-11-7
EINECS: 201-515-5
UN: 3077
ADR: 9,III
Merck Index: 12,07355

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR