1-Chloro-2,2-dimethylpropane ≥99%

Synonyms: Chloro-1-diméthyl-2,2-propane, neo-Pentylchloride

274496-5G 274496-25G
220046-918EA 71.08 USD
220046-918 220046-917
1-Chloro-2,2-dimethylpropane ≥99%
1-Chloro-2,2-dimethylpropane
Formula: C₅H₁₁Cl
MW: 106.6 g/mol
Boiling Pt: 84 °C (1013 hPa)
Melting Pt: –20 °C
Density: 0.86 g/cm³ (20 °C)
MDL Number: MFCD00000941
CAS Number: 753-89-9
EINECS: 212-040-8
UN: 1107
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR