Methyl-3,4,5-trihydroxybenzoate ≥98%

Synonyms: Methyl gallate, Gallic acid methyl ester, Methyl 3,4,5-trihydroxybenzoate

274194-5G 274194-100G
220048-475EA 53.62 USD
220048-475 220050-543
Methyl-3,4,5-trihydroxybenzoate ≥98%
Methyl-3,4,5-trihydroxybenzoate
Formula: C₈H₈O₅
MW: 184.15 g/mol
Boiling Pt: 450 °C (1013 hPa)
Melting Pt: 200…203 °C
MDL Number: MFCD00002194
CAS Number: 99-24-1
EINECS: 202-741-7
Merck Index: 13,04366

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR