1,6-Dihydroxynaphthalene

Warning

Synonyms: Naphthalene-1,6-diol

274127-250G 274127-50G
220050-533EA 486.46 USD
220050-533 220050-534
1,6-Dihydroxynaphthalene
1,6-Dihydroxynaphthalene
Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 135.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00003981
CAS Number: 575-44-0
EINECS: 209-386-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR