2-Chlorobenzoxazole

Synonyms: 2-Chloro-1,3-benzoxazole

274089-1G 274089-10G
220050-531EA 69.92 USD
220050-531 220050-530
2-Chlorobenzoxazole
2-Chlorobenzoxazole
Formula: C₇H₄ClNO
MW: 153.57 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: 45 °C
Density: 1.3453 g/cm³ (18 °C)
MDL Number: MFCD00005766
CAS Number: 615-18-9
EINECS: 210-414-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR