(+)-Cinchonine

Synonyms: Cinchonine, (S)-Quinolin-4-yl((1S,2R,4S,5R)-5-vinylquinuclidin-2-yl)methanol

27370-25G 27370-100G
220048-897EA 65.05 USD
220048-897 220048-896
(+)-Cinchonine
(+)-Cinchonine
Formula: C₁₉H₂₂N₂O
MW: 294.4 g/mol
Melting Pt: 261 - 264 °C
Density: 1.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064372
CAS Number: 118-10-5
EINECS: 204-234-6
Merck Index: 13,02308

Order Now

Specification Test Results

Assay 98.0% (NT)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR