2-Chloro-p-cresol ≥97%

Warning

Synonyms: 1-Chloro-2-hydroxy-5-methylbenzene, 2-Chloro-4-methylphenol

273198-25G 273198-5G
220050-512EA 282.15 USD
220050-512 220050-513
2-Chloro-p-cresol ≥97%
2-Chloro-p-cresol
Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 198 - 203 °C (1013 hPa)
Melting Pt: 76 °C
Density: 1.211 g/cm³ (25 °C)
MDL Number: MFCD00070639
CAS Number: 6640-27-3
EINECS: 229-656-8
UN: 2669
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR