2,4,6-Triphenylaniline ≥97%

Synonyms: 5'-Phenyl-[1,1':3',1''-terphenyl]-2'-amine

272515-5G 272515-1G
220050-419EA 365.08 USD
220050-419 220049-745
2,4,6-Triphenylaniline ≥97%
2,4,6-Triphenylaniline
Formula: C₂₄H₁₉N
MW: 321.42 g/mol
MDL Number: MFCD00012371
CAS Number: 6864-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR