ω-Chloro-2,4-difluoroacetophenone ≥98%

Synonyms: 2-Chloro-1-(2,4-difluorophenyl)ethanone, α-Chloro-2,4-difluoroacetophenone, 2-Chloro-2',4'-difluoroacetophenone

272507-5G 272507-25G
220049-744EA 140 USD
220049-744 220049-743
ω-Chloro-2,4-difluoroacetophenone ≥98%
ω-Chloro-2,4-difluoroacetophenone
Formula: C₈H₅ClF₂O
MW: 190.58 g/mol
Boiling Pt: 233 °C (1013 hPa)
Melting Pt: 47…48 °C
MDL Number: MFCD00013252
CAS Number: 51336-94-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR