1-(3-Aminopropyl)imidazole

272264-250G 272264-50G
220050-392EA 200.31 USD
220050-392 220050-393
1-(3-Aminopropyl)imidazole
1-(3-Aminopropyl)imidazole
Formula: C₆H₁₁N₃
MDL Number: MFCD00009819
CAS Number: 5036-48-6
EINECS: 225-730-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR