Tributylvinyltin

Danger

Synonyms: Tri-n-butylvinyltin, Tributylvinylstannane

271438-1G 271438-25G 271438-5G
220049-620EA 43.75 USD
220049-620 220049-621 220049-622
Tributylvinyltin
Tributylvinyltin
Formula: C₁₄H₃₀Sn
MW: 317.1 g/mol
Boiling Pt: 73…74 °C (0.1 mmHg)
Density: 1.085 g/cm³ (25 °C)
MDL Number: MFCD00009421
CAS Number: 7486-35-3
EINECS: 231-291-4
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR