4-tert-Butyl-1-(bromomethyl)benzene ≥97%

Danger

Synonyms: 1-(Bromomethyl)-4-(tert-butyl)benzene, 4-(tert-Butyl)benzylbromide

270768-25G 270768-5G
220050-158EA 146.46 USD
220050-158 220050-159
4-tert-Butyl-1-(bromomethyl)benzene ≥97%
4-tert-Butyl-1-(bromomethyl)benzene
Formula: C₁₁H₁₅Br
MW: 227.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000180
CAS Number: 18880-00-7
EINECS: 242-643-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR