3,4-Difluoroaniline ≥99%

Warning

270237-25G 270237-5G
220049-594EA 32.31 USD
220049-594 220049-595
3,4-Difluoroaniline ≥99%
3,4-Difluoroaniline
Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 22 °C
Density: 1.302 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007761
CAS Number: 3863-11-4
EINECS: 223-381-7
UN: 2941
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR