(±)-trans-1,2-Diaminocyclohexane ≥99%

Synonyms: (±)-trans-1,2-Cyclohexanediamine, trans-Cyclohexane-1,2-diamine

270016-250ML 270016-10ML 270016-50ML
220050-058EA 596.62 USD
220050-058 220050-057 220050-059
(±)-trans-1,2-Diaminocyclohexane ≥99%
(±)-trans-1,2-Diaminocyclohexane
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 187 °C (1013 hPa)
Melting Pt: 14.8 °C
Density: 0.951 g/cm³ (20 °C)
MDL Number: MFCD00063747
CAS Number: 1121-22-8

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR