4-tert-Butylbenzenesulfonyl chloride ≥98%

Synonyms: 4-tert-Butylbenzene-1-sulphonyl chloride

269670-50G 269670-10G
220050-027EA 369.38 USD
220050-027 220050-026
4-tert-Butylbenzenesulfonyl chloride ≥98%
4-tert-Butylbenzenesulphonyl chloride
Formula: C₁₀H₁₃ClO₂S
MW: 232.73 g/mol
Boiling Pt: 165 °C (18 mmHg)
Melting Pt: 83 °C
MDL Number: MFCD00007449
CAS Number: 15084-51-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR