N-Methyl-4-nitroaniline ≥97%

269638-25G 269638-100G
220050-025EA 53.31 USD
220050-025 220050-024
N-Methyl-4-nitroaniline ≥97%
N-Methyl-4-nitroaniline
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 291 °C (1013 hPa)
Melting Pt: 148…149 °C
Density: 1.201 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007305
CAS Number: 100-15-2
EINECS: 202-823-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR