Devarda's alloy

Synonyms: Copper aluminium zinc alloy (50:45:5 w%), Devarda's metal

269484-500G 269484-100G
220050-017EA 270.77 USD
220050-017 220050-016
Devarda's alloy
Devarda's alloy
Boiling Pt: 906 °C (1013 hPa)
Melting Pt: 490…560 °C
Density: 5.79 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00198176
CAS Number: 8049-11-4
UN: 3089
ADR: 4.1,II
Merck Index: 13,02958

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR