Diallyl phthalate ≥97%

Warning

Synonyms: Phthalic acid diallyl ester

269379-1L 269379-250ML
220050-006EA 110.92 USD
220050-006 220050-007
Diallyl phthalate ≥97%
Diallyl phthalate
Formula: C₁₄H₁₄O₄
MW: 246.26 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: -70 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 165 °C
Storage Temperature: Ambient
MDL Number: MFCD00008646
CAS Number: 131-17-9
EINECS: 205-016-3
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR