Methanesulfonic anhydride ≥97%

Danger

269190-25G 269190-5G
220049-992EA 88.74 USD
220049-992 220049-993
Methanesulfonic anhydride ≥97%
Methanesulphonic anhydride
Formula: C₂H₆O₅S₂
MW: 174.2 g/mol
Melting Pt: 63 - 66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007556
CAS Number: 7143-01-3
EINECS: 230-442-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR