1,3-Benzenedithiol ≥99%

Synonyms: Benzene-1,3-dithiol

269093-1G 269093-5G
220049-729EA 128.15 USD
220049-729 220049-730
1,3-Benzenedithiol ≥99%
1,3-Benzenedithiol
Formula: C₆H₆S₂
MW: 142.25 g/mol
Boiling Pt: 62 °C (0.2 mmHg)
Melting Pt: 27 °C
MDL Number: MFCD00004842
CAS Number: 626-04-0
EINECS: 210-925-3
UN: 3335
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR