1-Amino-2,2,2-trifluoroethane ≥99.5%

Synonyms: 2,2,2-Trifluoroethylamine

269042-5G 269042-1G
220049-726EA 172.31 USD
220049-726 220049-725
1-Amino-2,2,2-trifluoroethane ≥99.5%
1-Amino-2,2,2-trifluoroethane
Formula: C₂H₄F₃N
MW: 99.06 g/mol
Boiling Pt: 35.85 °C (1013 hPa)
Density: 1.2452 g/cm³ (25 °C)
MDL Number: MFCD00008132
CAS Number: 753-90-2
EINECS: 212-041-3
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR