4-Pentyn-2-ol ≥98%

Danger

Synonyms: 1-Pentyn-4-ol

268992-5G 268992-25G
220049-723EA 62.14 USD
220049-723 220049-722
4-Pentyn-2-ol ≥98%
4-Pentyn-2-ol
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 126 °C (1013 hPa)
Density: 0.894 g/cm³ (17 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004555
CAS Number: 2117-11-5
EINECS: 218-321-1
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR