2-Isopropylaniline ≥97%

Warning

268887-5ML 268887-100ML
220049-678EA 50.08 USD
220049-678 220049-677
2-Isopropylaniline ≥97%
2-Isopropylaniline
Formula: C₉H₁₃N
MW: 135.21 g/mol
MDL Number: MFCD00007720
CAS Number: 643-28-7
EINECS: 211-397-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR