4-Isopropylbenzoic acid ≥98%

Warning

268402-25G 268402-5G
220049-958EA 147.54 USD
220049-958 220049-959
4-Isopropylbenzoic acid ≥98%
4-Isopropylbenzoic acid
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 115…117 °C
Density: 1.162 g/cm³ (20 °C)
MDL Number: MFCD00002564
CAS Number: 536-66-3
EINECS: 208-642-5
Merck Index: 12,02684

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR