Diethyl methylphosphonate

Warning

268119-5G 268119-25G
220049-949EA 96.38 USD
220049-949 220049-494
Diethyl methylphosphonate
Diethyl methylphosphonate
Formula: C₅H₁₃O₃P
MW: 152.13 g/mol
Boiling Pt: 190 - 194 °C (1013 hPa)
Density: 1.0406 g/cm³ (30 °C)
MDL Number: MFCD00009813
CAS Number: 683-08-9
EINECS: 211-667-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR