Molybdenum(VI) oxide ACS

Warning

Synonyms: Molybdenum trioxide

267856-500G 267856-100G
220049-941EA 284.31 USD
220049-941 220049-940
Molybdenum(VI) oxide ACS
Molybdenum(VI) oxide
Formula: MoO₃
MW: 143.94 g/mol
Boiling Pt: 1155 °C (1013 hPa)
Melting Pt: 795 °C
Density: 4.69 g/cm³ (20 °C)
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1,III
Merck Index: 13,06261

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR