Cobalt ≥99.5%

Danger

266655-500G 266655-100G
220049-526EA 261 USD
220049-526 220049-525
Cobalt ≥99.5%
Cobalt
Formula: Co
Boiling Pt: 2870 °C (1013 hPa)
Melting Pt: 1495 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now

Specification Test Results

Assay 99.5% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR