Cobalt ≥99.9%

Danger

266647-250G 266647-50G
220049-523EA 360 USD
220049-523 220049-524
Cobalt ≥99.9%
Cobalt
Formula: Co
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
UN: 3089
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR