2,4-Difluorobenzoyl chloride ≥98%

265292-25G 265292-5G
220049-659EA 57.92 USD
220049-659 220048-961
2,4-Difluorobenzoyl chloride ≥98%
2,4-Difluorobenzoyl chloride
Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Boiling Pt: 68 °C (1013 hPa)
Density: 1.42 g/cm³ (20 °C)
MDL Number: MFCD00000658
CAS Number: 72482-64-5
EINECS: 276-690-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR