2,5-Difluorobenzaldehyde ≥98%

Warning

265187-1G 265187-5G 265187-25G
220049-655EA 34.03 USD
220049-655 220049-657 220049-656
2,5-Difluorobenzaldehyde ≥98%
2,5-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 65 - 66 °C (20 -rr)
Density: 1.3 g/cm³ (20 °C)
MDL Number: MFCD00010327
CAS Number: 2646-90-4
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR