2,4-Difluorobenzaldehyde ≥98%

Danger

265179-5G 265179-25G
220049-654EA 43.08 USD
220049-654 220049-653
2,4-Difluorobenzaldehyde ≥98%
2,4-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 82 °C (42 -rr)
Density: 1.299 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010326
CAS Number: 1550-35-2
EINECS: 216-287-2
UN: 1989
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR