3,4-Difluorobenzaldehyde ≥97%

265160-5G 265160-25G
220049-652EA 49.54 USD
220049-652 220049-651
3,4-Difluorobenzaldehyde ≥97%
3,4-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 53 °C (1013 hPa)
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010328
CAS Number: 34036-07-2
EINECS: 422-180-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR