2,6-Difluorobenzaldehyde ≥98%

265152-25G 265152-5G
220049-649EA 131.38 USD
220049-649 220049-650
2,6-Difluorobenzaldehyde ≥98%
2,6-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 194 °C (1013 hPa)
Melting Pt: 15 - 17 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010293
CAS Number: 437-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR