(2,6-Difluorophenyl)acetic acid ≥97%

Warning

Synonyms: 2,6-Difluorophenylacetic acid

264482-5G 264482-1G
220049-439EA 45.46 USD
220049-439 220049-438
(2,6-Difluorophenyl)acetic acid ≥97%
(2,6-Difluorophenyl)acetic acid
Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
Boiling Pt: 259 °C (1013 hPa)
Melting Pt: 100…102 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010001
CAS Number: 85068-28-6
EINECS: 285-289-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR