α-Bromo-3,4-difluorotoluene ≥98%

Synonyms: 3,4-Difluorobenzyl bromide, 4-(Bromomethyl)-1,2-difluorobenzene

264458-5G 264458-25G
220048-806EA 35.43 USD
220048-806 220048-805
α-Bromo-3,4-difluorotoluene ≥98%
α-Bromo-3,4-difluorotoluene
Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 59…61 °C (3.5 mmHg)
Density: 1.61 g/cm³ (20 °C)
MDL Number: MFCD00009889
CAS Number: 85118-01-0
EINECS: 285-653-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR